Order Entry
Germany
ContactUsLinkComponent
Methyl 1-(tert-Butyl)-5-(2,4-dichlorophenyl)pyrazole-3-carboxylate
Methyl 1-(tert-Butyl)-5-(2,4-dichlorophenyl)pyrazole-3-carboxylate
Catalog # APOSOR470673-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 1-(tert-Butyl)-5-(2,4-dichlorophenyl)pyrazole-3-carboxylate
Catalog # APOSOR470673-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :327.206 g/mol
Storage Temperature:Ambient
CAS Number:2006277-42-3
Formula:C₁₅H₁₆Cl₂N₂O₂
MDL Number:MFCD26045230