Methyl 1-Methyl-7-indolecarboxylate
Catalog # APOSOR470584-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:189.211 g/mol
Storage Temperature:Ambient
CAS Number:167479-21-2
Formula:C₁₁H₁₁NO₂
MDL Number:MFCD12025398