3-(Boc-amino)-3-(4-tetrahydrothiopyranyl)-1-propanol
Catalog # APOSOR470349-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:275.408 g/mol
Storage Temperature:Ambient
CAS Number:898405-01-1
Formula:C₁₃H₂₅NO₃S
MDL Number:MFCD04115545