Order Entry
Germany
ContactUsLinkComponent
 
Diethyl 2-(6-Methoxy-3-pyridyl)malonate
Catalog # APOSOR470301-5G
Supplier:  Apollo Scientific
CAS Number:  
Diethyl 2-(6-Methoxy-3-pyridyl)malonate
Catalog # APOSOR470301-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :267.278 g/mol
Storage Temperature:Ambient
CAS Number:902130-84-1
Formula:C₁₃H₁₇NO₅
MDL Number:MFCD19442788