Order Entry
Germany
ContactUsLinkComponent
1-Methyl-6-nitrobenzimidazole
1-Methyl-6-nitrobenzimidazole
Catalog # APOSOR470255-5G
Supplier:  Apollo Scientific
CAS Number:  
1-Methyl-6-nitrobenzimidazole
Catalog # APOSOR470255-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :177.16 g/mol
Storage Temperature:Ambient
CAS Number:5381-79-3
Formula:C₈H₇N₃O₂
MDL Number:MFCD00962814