Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 4-Thiomorpholinobutyrate
Methyl 4-Thiomorpholinobutyrate
Methyl 4-Thiomorpholinobutyrate
APOSOR470246-5G
:
Apollo Scientific
:
Methyl 4-Thiomorpholinobutyrate
APOSOR470246-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
203.302 g/mol
:
Ambient
:
443796-04-1
:
C₉H₁₇NO₂S
:
MFCD09880002