(R)-2-(N-Cbz-3-pyrrolidinyl)-2-propanol
Catalog # APOSOR470160-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:263.332 g/mol
Storage Temperature:Ambient
CAS Number:1417789-56-0
Formula:C₁₅H₂₁NO₃
MDL Number:MFCD18071038