Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 5-amino-2-chloroisonicotinate
Methyl 5-amino-2-chloroisonicotinate 95%
Methyl 5-amino-2-chloroisonicotinate 95%
:
Apollo Scientific
Methyl 5-amino-2-chloroisonicotinate 95%
:
Apollo Scientific
:
1073182-59-8
:
:
C₇H₇ClN₂O₂
:
MFCD11848213
:
186.596 g/mol
:
GHS07
:
Ambient