Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 2-bromo-5-nitroisonicotinate
Methyl 2-bromo-5-nitroisonicotinate 99%
Methyl 2-bromo-5-nitroisonicotinate 99%
APOSOR46523-250MG
:
Apollo Scientific
:
Methyl 2-bromo-5-nitroisonicotinate 99%
APOSOR46523-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
261.03 g/mol
:
GHS07
:
Ambient
:
1361385-86-5
:
C₇H₅BrN₂O₄
:
MFCD21606315