
Piperidine-4-sulphonyl chloride, N-CBZ protected, Technical Grade
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:287953-54-2
:
Formula:C₁₃H₁₆ClNO₄S
MDL Number:MFCD05664659
:317.788 g/mol
Storage Temperature:Ambient
UN:1759
ADR:8,III
:GHS05