
1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2(1H)-one 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1002309-52-5
:
Formula:C₁₂H₁₈BNO₃
MDL Number:MFCD11044683
:235.087 g/mol
:GHS07
Storage Temperature:Ambient