Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1H-Pyrazol-3-amine hydrochloride
1H-Pyrazol-3-amine hydrochloride
1H-Pyrazol-3-amine hydrochloride
1H-Pyrazol-3-amine hydrochloride
Supplier:
Apollo Scientific
Certificates
Expand image
1H-Pyrazol-3-amine hydrochloride
Supplier:
Apollo Scientific
About this item
CAS Number:
34045-29-9
:
Formula:
C₃H₅N₃·ClH
MDL Number:
MFCD07367630
:
119.553 g/mol
:
GHS07
Storage Temperature:
Ambient