Order Entry
Germany
ContactUsLinkComponent
Methyl 4-azaindole-3-carboxylate 97%
Irritant
Methyl 4-azaindole-3-carboxylate 97%
Catalog # APOSOR41124-50MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl 4-azaindole-3-carboxylate 97%
Catalog # APOSOR41124-50MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :176.172 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:952800-39-4
Formula:C₉H₈N₂O₂
MDL Number:MFCD09864655