
Methyl 5-bromo-1H-pyrrole-3-carboxylate 97+%
Catalog # APOSOR41103-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:204.021 g/mol
Melting Pt:109…111 °C
:GHS07
Storage Temperature:Ambient
CAS Number:16420-39-6
Formula:C₆H₆BrNO₂
MDL Number:MFCD09841652