Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-2-iodobenzoic acid
4-Bromo-2-iodobenzoic acid 97%
4-Bromo-2-iodobenzoic acid 97%
:
Apollo Scientific
4-Bromo-2-iodobenzoic acid 97%
:
Apollo Scientific
:
1133123-02-0
:
:
C₇H₄BrIO₂
:
MFCD12767500
:
326.914 g/mol
:
183…186 °C
:
GHS07
:
Ambient