Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Aminomethyl)-4-methylpyridine hydrochloride
2-(Aminomethyl)-4-methylpyridine hydrochloride 95%
2-(Aminomethyl)-4-methylpyridine hydrochloride 95%
Irritant
2-(Aminomethyl)-4-methylpyridine hydrochloride 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-(Aminomethyl)-4-methylpyridine hydrochloride 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1257535-59-3
:
Formula:
C₇H₁₀N₂·ClH
MDL Number:
MFCD16628187
:
158.629 g/mol
:
GHS07
Storage Temperature:
Ambient