Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Chloro-5-nitrobenzyl alcohol
3-Chloro-5-nitrobenzyl alcohol
3-Chloro-5-nitrobenzyl alcohol
Irritant
3-Chloro-5-nitrobenzyl alcohol
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-Chloro-5-nitrobenzyl alcohol
Supplier:
Apollo Scientific
About this item
CAS Number:
79944-62-0
:
Formula:
C₇H₆ClNO₃
MDL Number:
MFCD08236779
:
187.58 g/mol
:
GHS07
Storage Temperature:
Ambient