Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-3-chloroiodobenzene
2-Bromo-3-chloroiodobenzene 95%
2-Bromo-3-chloroiodobenzene 95%
2-Bromo-3-chloroiodobenzene 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Bromo-3-chloroiodobenzene 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1369793-66-7
:
Formula:
C₆H₃BrClI
MDL Number:
MFCD28397823
:
317.349 g/mol
:
GHS07
Storage Temperature:
Ambient