Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Hydroxybenzo[b]furan
7-Hydroxybenzo[b]furan
7-Hydroxybenzo[b]furan
Irritant
7-Hydroxybenzo[b]furan
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
7-Hydroxybenzo[b]furan
Supplier:
Apollo Scientific
About this item
CAS Number:
4790-81-2
:
Formula:
C₈H₆O₂
MDL Number:
MFCD03788248
:
134.132 g/mol
Boiling Pt:
240 °C
Melting Pt:
42…43 °C
:
GHS07
Storage Temperature:
Refrigerator