Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-4-hydroxy-3-methoxybenzaldehyde
2-Chloro-4-hydroxy-3-methoxybenzaldehyde ≥98%
2-Chloro-4-hydroxy-3-methoxybenzaldehyde ≥98%
2-Chloro-4-hydroxy-3-methoxybenzaldehyde ≥98%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Chloro-4-hydroxy-3-methoxybenzaldehyde ≥98%
Supplier:
Apollo Scientific
About this item
CAS Number:
82668-20-0
:
Formula:
C₈H₇ClO₃
MDL Number:
MFCD11042341
:
186.592 g/mol
Storage Temperature:
Ambient