Order Entry
Germany
ContactUsLinkComponent
2-Chloro-1-(6-methoxypyridin-3-yl)ethanone ≥98%
2-Chloro-1-(6-methoxypyridin-3-yl)ethanone ≥98%
Catalog # APOSOR361738-100MG
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-1-(6-methoxypyridin-3-yl)ethanone ≥98%
Catalog # APOSOR361738-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :185.608 g/mol
Storage Temperature:Ambient
CAS Number:136591-96-3
Formula:C₈H₈ClNO₂
MDL Number:MFCD20635182