Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Methylpentan-2-amine hydrochloride
N-Methylpentan-2-amine hydrochloride ≥98%
N-Methylpentan-2-amine hydrochloride ≥98%
APOSOR361721-500MG
:
Apollo Scientific
:
N-Methylpentan-2-amine hydrochloride ≥98%
APOSOR361721-500MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
500 mg
Seq:
2725
:
:
137.651 g/mol
:
Ambient
:
130985-80-7
:
C₆H₁₅N·ClH
:
MFCD11099634