Order Entry
Germany
ContactUsLinkComponent
4-(Methoxycarbonyl)cyclohexene-1-boronic acid, pinacol ester 97%
4-(Methoxycarbonyl)cyclohexene-1-boronic acid, pinacol ester 97%
Catalog # APOSOR361328-250MG
Supplier:  Apollo Scientific
CAS Number:  
4-(Methoxycarbonyl)cyclohexene-1-boronic acid, pinacol ester 97%
Catalog # APOSOR361328-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :266.141 g/mol
Storage Temperature:Ambient
CAS Number:151075-20-6
Formula:C₁₄H₂₃BO₄
MDL Number:MFCD11520532