Order Entry
Germany
ContactUsLinkComponent
1-N-BOC-3-methyl-indazole-5-boronic acid, pinacol ester 97%
1-N-BOC-3-methyl-indazole-5-boronic acid, pinacol ester 97%
Catalog # APOSOR360108-250MG
Supplier:  Apollo Scientific
CAS Number:  
1-N-BOC-3-methyl-indazole-5-boronic acid, pinacol ester 97%
Catalog # APOSOR360108-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :358.24 g/mol
Storage Temperature:Ambient
CAS Number:864770-82-1
Formula:C₁₉H₂₇BN₂O₄
MDL Number:MFCD13195073