Order Entry
Germany
ContactUsLinkComponent
4-Acetyl-1H-pyrrole-2-carboxaldehyde ≥97%
4-Acetyl-1H-pyrrole-2-carboxaldehyde ≥97%
Catalog # APOSOR346429-250MG
Supplier:  Apollo Scientific
CAS Number:  
4-Acetyl-1H-pyrrole-2-carboxaldehyde ≥97%
Catalog # APOSOR346429-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :137.136 g/mol
Storage Temperature:Refrigerator
CAS Number:16168-92-6
Formula:C₇H₇NO₂
MDL Number:MFCD02179580