Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-quinoline-3-carboxylic acid amide
2-Amino-quinoline-3-carboxylic acid amide
2-Amino-quinoline-3-carboxylic acid amide
2-Amino-quinoline-3-carboxylic acid amide
Supplier:
Apollo Scientific
Certificates
Expand image
2-Amino-quinoline-3-carboxylic acid amide
Supplier:
Apollo Scientific
About this item
CAS Number:
31407-28-0
:
Formula:
C₁₀H₉N₃O
MDL Number:
MFCD06809614
:
187.198 g/mol
:
GHS07
Storage Temperature:
Ambient