tert-Butyl 9-cyano-3-azaspiro[5.5]undecane-3-carboxylate, tech
Catalog # APOSOR33588-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:278.39 g/mol
Storage Temperature:Ambient
CAS Number:1281872-59-0
Formula:C₁₆H₂₆N₂O₂
MDL Number:MFCD18426549