Order Entry
Germany
ContactUsLinkComponent
3-(Cyclohex-3-en-1-yl)-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid, tech
3-(Cyclohex-3-en-1-yl)-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid, tech
Catalog # APOSOR33407-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(Cyclohex-3-en-1-yl)-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid, tech
Catalog # APOSOR33407-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :285.338 g/mol
Storage Temperature:Ambient
CAS Number:866144-53-8
Formula:C₁₇H₁₉NO₃
MDL Number:MFCD03305894