2,4-Dimethylimidazo[1,5-a]pyrimidine-8-carbonitrile, tech
Catalog # APOSOR33371-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:172.187 g/mol
Storage Temperature:Ambient
CAS Number:55418-17-2
Formula:C₉H₈N₄
MDL Number:MFCD00975066