Order Entry
Germany
ContactUsLinkComponent
(2E)-1-{6-Chloro-2-methylimidazo[1,2-a]pyridin-3-yl}-3-(dimethylamino)prop-2-en-1-one, tech
(2E)-1-{6-Chloro-2-methylimidazo[1,2-a]pyridin-3-yl}-3-(dimethylamino)prop-2-en-1-one, tech
Catalog # APOSOR33183-1G
Supplier:  Apollo Scientific
CAS Number:  
(2E)-1-{6-Chloro-2-methylimidazo[1,2-a]pyridin-3-yl}-3-(dimethylamino)prop-2-en-1-one, tech
Catalog # APOSOR33183-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :263.723 g/mol
Storage Temperature:Ambient
CAS Number:900015-66-9
Formula:C₁₃H₁₄ClN₃O
MDL Number:MFCD06496193