2-[(E)-2-(Dimethylamino)ethenyl]-9,10-dihydroanthracene-9,10-dione, tech
Catalog # APOSOR32867-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:277.317 g/mol
Storage Temperature:Ambient
CAS Number:153813-68-4
Formula:C₁₈H₁₅NO₂
MDL Number:MFCD01315203