Order Entry
Germany
ContactUsLinkComponent
1-{6-Chloro-2-methylimidazo[1,2-a]pyridin-3-yl}ethan-1-one, tech
1-{6-Chloro-2-methylimidazo[1,2-a]pyridin-3-yl}ethan-1-one, tech
Catalog # APOSOR32671-1G
Supplier:  Apollo Scientific
CAS Number:  
1-{6-Chloro-2-methylimidazo[1,2-a]pyridin-3-yl}ethan-1-one, tech
Catalog # APOSOR32671-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :208.644 g/mol
Storage Temperature:Ambient
CAS Number:154877-60-8
Formula:C₁₀H₉ClN₂O
MDL Number:MFCD00973744