1-{2-Methylimidazo[1,2-a]pyridin-3-yl}-2-(morpholin-4-yl)ethan-1-one, tech
Catalog # APOSOR32670-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:259.304 g/mol
Storage Temperature:Ambient
CAS Number:383148-01-4
Formula:C₁₄H₁₇N₃O₂
MDL Number:MFCD03012040