Order Entry
Germany
ContactUsLinkComponent
5-Methyl-2-phenylpyrazolo[1,5-a]pyrimidin-7-ol 95%
5-Methyl-2-phenylpyrazolo[1,5-a]pyrimidin-7-ol 95%
Catalog # APOSOR32479-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Methyl-2-phenylpyrazolo[1,5-a]pyrimidin-7-ol 95%
Catalog # APOSOR32479-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :225.246 g/mol
Storage Temperature:Ambient
CAS Number:90019-55-9
Formula:C₁₃H₁₁N₃O
MDL Number:MFCD00634653