1-(3-Chlorophenyl)-3-hydroxy-3-methylurea, tech
Catalog # APOSOR32447-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:200.622 g/mol
Storage Temperature:Ambient
CAS Number:26816-97-7
Formula:C₈H₉ClN₂O₂
MDL Number:MFCD00459805