Order Entry
Germany
ContactUsLinkComponent
7-Phenylpyrazolo[1,5-a]pyrimidine-3-carbonitrile, tech
7-Phenylpyrazolo[1,5-a]pyrimidine-3-carbonitrile, tech
Catalog # APOSOR32381-1G
Supplier:  Apollo Scientific
CAS Number:  
7-Phenylpyrazolo[1,5-a]pyrimidine-3-carbonitrile, tech
Catalog # APOSOR32381-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :220.229 g/mol
Storage Temperature:Ambient
CAS Number:72860-56-1
Formula:C₁₃H₈N₄
MDL Number:MFCD00138880