Order Entry
Germany
ContactUsLinkComponent
SBAP (Succinimidyl 3-(bromoacetamido)propionate)
SBAP (Succinimidyl 3-(bromoacetamido)propionate)
Catalog # APOSOR322800-100MG
Supplier:  Apollo Scientific
CAS Number:  
SBAP (Succinimidyl 3-(bromoacetamido)propionate)
Catalog # APOSOR322800-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :307.098 g/mol
Storage Temperature:Ambient
CAS Number:57159-62-3
Formula:C₉H₁₁BrN₂O₅
MDL Number:MFCD01863457