tert-Butyl 6-chloro-2-iodo-1H-pyrrolo[3,2-c]pyridine-1-carboxylate
Catalog # APOSOR322773-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:378.593 g/mol
Storage Temperature:Ambient
CAS Number:1421933-27-8
Formula:C₁₂H₁₂ClIN₂O₂
MDL Number:MFCD23378567