Order Entry
Germany
ContactUsLinkComponent
2-Methoxy-4-methylaniline
2-Methoxy-4-methylaniline
Catalog # APOSOR322306-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Methoxy-4-methylaniline
Catalog # APOSOR322306-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :137.179 g/mol
Storage Temperature:Ambient
CAS Number:39538-68-6
Formula:C₈H₁₁NO
MDL Number:MFCD06408246