Methyl 1-[(4-methylphenyl)methyl]piperidine-4-carboxylate, tech
Catalog # APOSOR32228-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:247.333 g/mol
Storage Temperature:Ambient
CAS Number:477856-45-4
Formula:C₁₅H₂₁NO₂
MDL Number:MFCD02082486