Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tert-Butyl 3-cyanopiperazine-1-carboxylate
tert-Butyl 3-cyanopiperazine-1-carboxylate 97%
tert-Butyl 3-cyanopiperazine-1-carboxylate 97%
APOSOR321305-1G
:
Apollo Scientific
:
tert-Butyl 3-cyanopiperazine-1-carboxylate 97%
APOSOR321305-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
211.261 g/mol
:
GHS07
:
Ambient
:
859518-35-7
:
C₁₀H₁₇N₃O₂
:
MFCD09991479