Order Entry
Germany
ContactUsLinkComponent
 
tert-Butyl 4-(4-(2-hydroxyethyl)phenyl)piperidine-1-carboxylate 95%
  APOSOR321261-100MG
 :  
tert-Butyl 4-(4-(2-hydroxyethyl)phenyl)piperidine-1-carboxylate 95%
  APOSOR321261-100MG
 :  

 

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

 

 

 :
 :305.412 g/mol
 :GHS07
 :Refrigerator
 :1853217-74-9
 :C₁₈H₂₇NO₃
 :MFCD30099265