Order Entry
Germany
ContactUsLinkComponent
tert-Butyl 4-(2-(2-hydroxyethyl)phenyl)piperidine-1-carboxylate 97%
tert-Butyl 4-(2-(2-hydroxyethyl)phenyl)piperidine-1-carboxylate 97%
Catalog # APOSOR321260-100MG
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl 4-(2-(2-hydroxyethyl)phenyl)piperidine-1-carboxylate 97%
Catalog # APOSOR321260-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :305.412 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:170837-78-2
Formula:C₁₈H₂₇NO₃
MDL Number:MFCD30099263