tert-Butyl (S)-2-ethylpiperidine-1-carboxylate 97%
Catalog # APOSOR321206-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:213.317 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1853230-28-0
Formula:C₁₂H₂₃NO₂
MDL Number:MFCD28355903