5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H,2H,3H-pyrrolo[2,3-b]pyridin-2-one
Catalog # APOSOR317332-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:260.097 g/mol
Storage Temperature:Ambient
CAS Number:1207623-97-9
Formula:C₁₃H₁₇BN₂O₃
MDL Number:MFCD16995984