3-[(Methoxymethylamino)carbonyl]-1-azetidinecarboxylic acid, 1,1-dimethylethyl ester
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:820971-67-3
:
Formula:C₁₁H₂₀N₂O₄
MDL Number:MFCD11975576
:244.287 g/mol
Storage Temperature:Ambient