Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(Cyclopropyl)thiophene-2-carboxaldehyde
3-(Cyclopropyl)thiophene-2-carboxaldehyde 95%
3-(Cyclopropyl)thiophene-2-carboxaldehyde 95%
3-(Cyclopropyl)thiophene-2-carboxaldehyde 95%
Supplier:
Apollo Scientific
Certificates
Expand image
3-(Cyclopropyl)thiophene-2-carboxaldehyde 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
29481-31-0
:
Formula:
C₈H₈OS
MDL Number:
MFCD22070104
:
152.214 g/mol
Storage Temperature:
Refrigerator