tert-Butyl 6-methoxy-7-nitro-1,2,3,4-tetrahydroisoquinoline-2-carboxylate
Catalog # APOSOR310935-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:308.33 g/mol
Storage Temperature:Ambient
CAS Number:921224-62-6
Formula:C₁₅H₂₀N₂O₅
MDL Number:MFCD28101536