Order Entry
Germany
ContactUsLinkComponent
(2S)-3-[(4-Methoxyphenyl)methoxy]-2-methylpropan-1-ol
Irritant
(2S)-3-[(4-Methoxyphenyl)methoxy]-2-methylpropan-1-ol
Catalog # APOSOR310541-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(2S)-3-[(4-Methoxyphenyl)methoxy]-2-methylpropan-1-ol
Catalog # APOSOR310541-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :210.27 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:160238-45-9
Formula:C₁₂H₁₈O₃
MDL Number:MFCD27957342