4-Hydroxy-7-methoxy-2-propylquinoline
Catalog # APOSOR309306-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:217.264 g/mol
Storage Temperature:Ambient
CAS Number:1070879-90-1
Formula:C₁₃H₁₅NO₂
MDL Number:MFCD11505253